Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | rno-miR-17-1-3p |
---|---|
miRBase Accession Number: | MI0000845 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGCAGUGAAGGCACUUGUGG |
Species: | Rat |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCAGUGAAGGCACUUGUGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-17-1-3p | MIMAT0004710 |