Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | rno-miR-18a-3p |
---|---|
miRBase Accession Number: | MI0000846 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGCCCUAAGUGCUCCUUCU |
Species: | Rat, Xenopus tropicalis |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCCCUAAGUGCUCCUUCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-18a-3p | MIMAT0017095 |
Xenopus tropicalis | xtr-miR-18a-3p | MIMAT0005462 |