Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-19a-3p |
---|---|
miRBase Accession Number: | MI0000073 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UGUGCAAAUCUAUGCAAAACUGA |
Species: | Human, Mouse, Rat, Ateles geoffroyi, Bovine, Callithrix jacchus, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Fugu rubripes, Gadus morhua, Goat, Gorilla gorilla, Guinea pig, Horse, Lagothrix lagotricha, Lemur catta, Macaca nemestrina, Monodelphis domestica, Nomascus leucogenys, Ophiophagus hannah, Ornithorhynchus anatinus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rabbit, Rhesus monkey, Saguinus labiatus, Tetraodon nigroviridis, Tupaia chinensis, Xenopus tropicalis, Zebrafish |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000011, mir-19 |
Mature miRNA Sequence: | UGUGCAAAUCUAUGCAAAACUGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-19a-3p | MIMAT0000073 |
Mouse | mmu-miR-19a-3p | MIMAT0000651 |
Rat | rno-miR-19a-3p | MIMAT0000789 |
Ateles geoffroyi | age-miR-19a | MIMAT0002675 |
Bovine | bta-miR-19a | MIMAT0004336 |
Callithrix jacchus | cja-miR-19a | MIMAT0049329 |
View More Rows | ||
Chicken | gga-miR-19a-3p | MIMAT0001112 |
Columba livia | cli-miR-19a-3p | MIMAT0038428 |
Cricetulus griseus | cgr-miR-19a | MIMAT0023839 |
Dasypus novemcinctus | dno-miR-19a-3p | MIMAT0047516 |
Dog | cfa-miR-19a | MIMAT0006650 |
Fugu rubripes | fru-miR-19a | MIMAT0002920 |
Gadus morhua | gmo-miR-19a-3p | MIMAT0044303 |
Goat | chi-miR-19a | MIMAT0036042 |
Gorilla gorilla | ggo-miR-19a | MIMAT0002661 |
Guinea pig | cpo-miR-19a-3p | MIMAT0046864 |
Horse | eca-miR-19a | MIMAT0013086 |
Lagothrix lagotricha | lla-miR-19a | MIMAT0002717 |
Lemur catta | lca-miR-19a | MIMAT0002668 |
Macaca nemestrina | mne-miR-19a | MIMAT0002724 |
Monodelphis domestica | mdo-miR-19a-3p | MIMAT0004168 |
Nomascus leucogenys | nle-miR-19a | MIMAT0049454 |
Ophiophagus hannah | oha-miR-19a-3p | MIMAT0036796 |
Ornithorhynchus anatinus | oan-miR-19a-3p | MIMAT0007148 |
Pan paniscus | ppa-miR-19a | MIMAT0002682 |
Pan troglodytes | ptr-miR-19a | MIMAT0002696 |
Pig | ssc-miR-19a | MIMAT0002128 |
Pongo pygmaeus | ppy-miR-19a | MIMAT0002689 |
Python bivittatus | pbv-miR-19a-3p | MIMAT0038844 |
Rabbit | ocu-miR-19a-3p | MIMAT0048116 |
Rhesus monkey | mml-miR-19a-3p | MIMAT0002703 |
Saguinus labiatus | sla-miR-19a | MIMAT0002710 |
Tetraodon nigroviridis | tni-miR-19a | MIMAT0002921 |
Tupaia chinensis | tch-miR-19a-3p | MIMAT0036603 |
Xenopus tropicalis | xtr-miR-19a | MIMAT0003566 |
Zebrafish | dre-miR-19a-3p | MIMAT0001782 |