Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | rno-miR-19b-2-5p |
---|---|
miRBase Accession Number: | MI0000848 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGUUUUGCAGAUUUGCAGUUCAG |
Species: | Rat |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000011, mir-19 |
Mature miRNA Sequence: | AGUUUUGCAGAUUUGCAGUUCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-19b-2-5p | MIMAT0017097 |