- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: ADAMTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014243.2 | NP_055058.2 | 15 | 2142 | ||
XM_011532421.2 | XP_011530723.1 | 15 | 2174 | |||
XM_011532422.2 | XP_011530724.1 | 15 | 2673 | |||
GenBank mRNA | AB002364.1 | - | 15 | 2094 | ||
BC130287.1 | - | 15 | 2139 | 40146318 | ||
BC132735.1 | - | 15 | 2139 | 40146757 |