Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: INHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002191.3 | NP_002182.1 | 2 | 1190 | ||
GenBank mRNA | BC006391.2 | - | 2 | 1127 | 4126990 | |
BT006954.1 | - | 2 | 1047 | |||
M13144.1 | - | 2 | 1004 | |||
M13981.1 | - | 2 | 1191 |