Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272101.1 | NP_001259030.1 | 7 | 1255 | ||
NM_002429.5 | NP_002420.1 | 9 | 1523 | |||
NR_073606.1 | 9 | 1600 | ||||
XM_006719401.3 | XP_006719464.1 | 7 | 1164 | |||
XM_011538359.1 | XP_011536661.1 | 8 | 1263 | |||
XM_017019309.1 | XP_016874798.1 | 8 | 1381 | |||
GenBank mRNA | AK225939.1 | - | 6 | 837 | ||
AK303202.1 | - | 7 | 1208 | |||
BC050368.1 | - | 9 | 1462 | 5761074 | ||
U37791.1 | - | 9 | 1451 | |||
U38320.1 | - | 3 | 2051 | |||
U38321.1 | - | 9 | 1446 | |||
U38322.1 | - | 5 | 1257 | |||
U38431.1 | - | 9 | 1521 | |||
X92521.1 | - | 9 | 1471 | |||
Y08622.1 | - | 9 | 1470 |