Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002424.2 | NP_002415.1 | 1, 2 | 209 | ||
GenBank mRNA | AK292659.1 | - | 1, 2 | 209 | ||
AK303511.1 | - | 2, 3 | 157 | |||
AK313374.1 | - | 1, 2 | 209 | |||
AL597614.1 | - | 1, 2 | 209 | |||
BC074988.2 | - | 1, 2 | 148 | 30915520 | ||
BC074989.2 | - | 1, 2 | 148 | 30915305 | ||
J05556.1 | - | 1, 2 | 181 |