Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TRIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003141.3 | NP_003132.2 | 4 | 658 | ||
GenBank mRNA | AY742713.1 | - | 3 | 545 | ||
BC010861.1 | - | 4 | 629 | 3900540 | ||
M34551.1 | - | 3 | 583 | |||
M62800.1 | - | 4 | 630 |