Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306089.1 | NP_001293018.1 | 2, 3 | 207 | ||
NM_007345.3 | NP_031371.3 | 2, 3 | 398 | |||
XM_005266762.3 | XP_005266819.1 | 2, 3 | 550 | |||
XM_011526165.2 | XP_011524467.1 | 3, 4 | 552 | |||
XM_011526168.2 | XP_011524470.1 | 2, 3 | 264 | |||
XM_011526169.2 | XP_011524471.1 | 2, 3 | 263 | |||
GenBank mRNA | AF085243.1 | - | 2, 3 | 398 | ||
AF085244.1 | - | 2, 3 | 398 | |||
BC140859.1 | - | 2, 3 | 229 | 9021727 | ||
BX648028.1 | - | 2, 3 | 207 |