Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KLF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138693.3 | NP_619638.2 | 1 | 844 | ||
GenBank mRNA | AF490374.1 | - | 1 | 844 | ||
DQ534757.1 | - | 1 | 918 |