Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293312.1 | NP_001280241.1 | 6 | 674 | ||
NM_001349.3 | NP_001340.2 | 7 | 732 | |||
XM_017003489.1 | XP_016858978.1 | 7 | 703 | |||
GenBank mRNA | AK129521.1 | - | 7 | 674 | ||
AK222476.1 | - | 7 | 562 | |||
AK290607.1 | - | 7 | 589 | |||
AY762100.1 | - | 7 | 551 | |||
BC000629.2 | - | 7 | 565 | 3344322 | ||
BC107749.1 | - | 7 | 545 | 6709590 | ||
BI753718.1 | - | 7 | 704 | |||
BT006710.1 | - | 7 | 516 | |||
CR749809.1 | - | 6 | 482 | |||
J05032.1 | - | 7 | 606 |