Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002472.2 | NP_002463.2 | 3 | 278 | ||
GenBank mRNA | AK303395.1 | - | 3 | 278 | ||
DB142109.1 | - | 3 | 278 | |||
M36769.1 | - | 2 | 256 | |||
Z38133.1 | - | 2 | 256 |