Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PADI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012387.2 | NP_036519.2 | 8, 9 | 957 | ||
XM_011541150.1 | XP_011539452.1 | 6, 7 | 769 | |||
XM_011541151.1 | XP_011539453.1 | 8, 9 | 957 | |||
XM_011541152.1 | XP_011539454.1 | 5, 6 | 489 | |||
XM_011541153.1 | XP_011539455.1 | 8, 9 | 957 | |||
XM_011541154.2 | XP_011539456.1 | 8, 9 | 957 | |||
XM_011541155.1 | XP_011539457.1 | 8, 9 | 957 | |||
XM_011541156.1 | XP_011539458.1 | 8, 9 | 957 | |||
XM_011541157.1 | XP_011539459.1 | 5, 6 | 436 | |||
GenBank mRNA | AB017919.1 | - | 8, 9 | 955 | ||
AK290507.1 | - | 8, 9 | 955 | |||
AK309637.1 | - | 8, 9 | 949 | |||
AK314839.1 | - | 8, 9 | 957 | |||
BC025718.1 | - | 8, 9 | 941 | 5228777 |