Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZBED5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143667.1 | NP_001137139.1 | 3 | 2504 | ||
NM_021211.3 | NP_067034.2 | 3 | 2536 | |||
GenBank mRNA | AF205600.1 | - | 3 | 2505 | ||
AK025407.1 | - | 1 | 1763 | |||
AK025829.1 | - | 1 | 2016 | |||
AK315033.1 | - | 3 | 2468 | |||
BC047754.1 | - | 3 | 2503 | 6471771 |