Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003655.2 | NP_003646.2 | 5 | 1094 | ||
XM_011525399.2 | XP_011523701.1 | 3 | 2603 | |||
GenBank mRNA | AF013956.1 | - | 5 | 1091 | ||
EU439707.1 | - | 5 | 1126 | |||
KU178423.1 | - | 5 | 931 |