Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PSKH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006742.2 | NP_006733.1 | 3 | 1965 | ||
GenBank mRNA | AJ272212.1 | - | 3 | 1923 | ||
BC021584.1 | - | 1 | 480 | |||
BC037560.1 | - | 1 | 835 | |||
BC062616.1 | - | 3 | 1936 | 5188431 |