Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286703.1 | NP_001273632.1 | 3 | 233 | ||
NM_001286704.1 | NP_001273633.1 | 3 | 411 | |||
NM_001286705.1 | NP_001273634.1 | 3 | 239 | |||
NM_001286706.1 | NP_001273635.1 | 3 | 239 | |||
NM_016617.3 | NP_057701.1 | 3 | 239 | |||
NR_104584.1 | 3 | 239 | ||||
NR_104585.1 | 3 | 239 | ||||
GenBank mRNA | AB154404.1 | - | 2 | 90 | ||
AF208844.1 | - | 3 | 129 | |||
AK095490.1 | - | 3 | 159 | |||
AK307111.1 | - | 3 | 159 | |||
AK309914.1 | - | 3 | 125 | |||
AL515797.3 | - | 2 | 91 | |||
BC005193.1 | - | 3 | 156 | 3829206 | ||
BG533062.1 | - | 3 | 332 | |||
BG540094.1 | - | 3 | 127 | |||
BG717690.1 | - | 3 | 161 | |||
BI458294.1 | - | 3 | 210 | |||
BI761948.1 | - | 3 | 116 | |||
BP277186.1 | - | 3 | 239 | |||
BU589105.1 | - | 3 | 155 | |||
CR457189.1 | - | 2 | 90 | |||
DA448931.1 | - | 3 | 152 | |||
DA483023.1 | - | 3 | 159 | |||
Z70222.1 | - | 3 | 130 |