Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LURAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203403.1 | NP_981948.1 | 1 | 999 | ||
XM_005251443.2 | XP_005251500.1 | 1 | 1022 | |||
GenBank mRNA | AB073606.1 | - | 1 | 1001 | ||
AK074752.1 | - | 1 | 480 | |||
AK095824.1 | - | 1 | 998 | |||
BC036923.1 | - | 1 | 974 | 5228515 | ||
BM739457.1 | - | 1 | 215 |