Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011436.3 | NP_035566.2 | 9 | 1464 | ||
GenBank mRNA | AB015790.1 | - | 9 | 1288 | ||
AF031816.1 | - | 742 | ||||
AK147303.1 | - | 9 | 1465 | |||
AK155045.1 | - | 9 | 1304 |