Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Skint10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177668.2 | NP_808336.1 | 7 | 1302 | ||
GenBank mRNA | AK037152.1 | - | 7 | 1267 | ||
BB749775.1 | - | 1 | 13 |