Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Armc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166138.1 | NP_001159610.1 | 3, 4 | 445 | ||
NM_028768.3 | NP_083044.2 | 3, 4 | 445 | |||
GenBank mRNA | AK004793.2 | - | 3, 4 | 446 | ||
AK078280.1 | - | 3, 4 | 196 | |||
AK083094.1 | - | 2, 3 | 137 | |||
BC038866.1 | - | 3, 4 | 403 | 1230321 |