Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 2210010C04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023333.4 | NP_075822.3 | 3 | 313 | ||
GenBank mRNA | AK007773.1 | - | 3 | 311 | ||
AK008460.1 | - | 3 | 314 | |||
AK008695.1 | - | 3 | 312 | |||
BC061093.1 | - | 3 | 314 | 30306963 |