Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tcf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253862.1 | NP_001240791.1 | 3 | 305 | ||
NM_001253865.1 | NP_001240794.1 | 3 | 362 | |||
NM_011544.3 | NP_035674.2 | 3 | 362 | |||
GenBank mRNA | AK035173.1 | - | 1 | 754 | ||
AK134037.1 | - | 3 | 306 | |||
AK143246.1 | - | 3 | 365 | |||
AK169800.1 | - | 3 | 391 | |||
BC037097.1 | - | 3 | 350 | 5345693 | ||
BY739497.1 | - | 3 | 363 | |||
M97635.1 | - | 3 | 333 | |||
X64840.1 | - | 3 | 162 |