Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Angptl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011923.4 | NP_036053.2 | 4 | 1557 | ||
GenBank mRNA | AF125176.1 | - | 3 | 1044 | ||
AK037265.1 | - | 4 | 1450 | |||
AK155464.1 | - | 4 | 1558 | |||
AK205240.1 | - | 2 | 123 | |||
AK214337.1 | - | 2 | 123 | |||
BC138609.1 | - | 4 | 1363 | 8861629 | ||
BC138610.1 | - | 4 | 1363 | 8861630 |