Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nphs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130828.2 | NP_570841.2 | 3, 4 | 514 | ||
GenBank mRNA | AB091379.1 | - | 3, 4 | 453 | ||
AF309631.1 | - | 2, 3 | 192 | |||
AF400653.1 | - | 3, 4 | 234 | |||
AY039651.1 | - | 3, 4 | 514 | |||
BC098649.1 | - | 3, 4 | 496 | 7127895 |