Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031640.1 | NP_113828.1 | 3 | 284 | ||
XM_008765417.1 | XP_008763639.1 | 2 | 385 | |||
GenBank mRNA | AF097723.1 | - | 3 | 284 | ||
AF131077.1 | - | 2 | 226 | |||
BC070884.1 | - | 2 | 245 | 7098277 | ||
FQ210652.1 | - | 1 | 24 |