Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem106c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008358.1 | NP_001008359.1 | 5 | 515 | ||
XM_006242326.2 | XP_006242388.1 | 5 | 703 | |||
GenBank mRNA | BC086606.1 | - | 5 | 515 | 7316815 |