Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nlrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004142.2 | NP_001004142.2 | 3 | 1286 | ||
GenBank mRNA | AK157034.1 | - | 3 | 1287 | ||
AY355339.1 | - | 2 | 1068 | |||
DQ117601.1 | - | 2 | 1227 | |||
KC539856.1 | - | 2 | 1068 | |||
KC539857.1 | - | 2 | 1068 | |||
KC539858.1 | - | 2 | 1068 | |||
KC539859.1 | - | 2 | 1068 | |||
KC539860.1 | - | 2 | 1068 | |||
KC539861.1 | - | 2 | 1068 |