Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MAP3K11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002419.3 | NP_002410.1 | 1 | 861 | ||
GenBank mRNA | AB209655.1 | - | 1 | 848 | ||
AK092015.1 | - | 1 | 2006 | |||
AK222781.1 | - | 1 | 854 | |||
AK315557.1 | - | 1 | 833 | |||
BC011263.2 | - | 1 | 849 | 4215281 | ||
BC064543.1 | - | 1 | 820 | 5733866 | ||
L32976.1 | - | 1 | 849 | |||
U07747.1 | - | 1 | 818 |