Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320044.1 | NP_001306973.1 | 9 | 1290 | ||
NM_012235.3 | NP_036367.2 | 12 | 1775 | |||
XM_005264967.1 | XP_005265024.1 | 12 | 1702 | |||
XM_005264968.1 | XP_005265025.1 | 9 | 1444 | |||
XM_005264971.1 | XP_005265028.1 | 6 | 855 | |||
XM_011533501.1 | XP_011531803.1 | 12 | 1802 | |||
XM_011533502.2 | XP_011531804.1 | 4 | 451 | |||
XM_017005918.1 | XP_016861407.1 | 12 | 1789 | |||
XM_017005919.1 | XP_016861408.1 | 12 | 1722 | |||
XM_017005920.1 | XP_016861409.1 | 12 | 1722 | |||
XM_017005921.1 | XP_016861410.1 | 6 | 855 | |||
GenBank mRNA | AK027402.1 | - | 6 | 768 | ||
AK291484.1 | - | 7 | 968 | |||
AK300311.1 | - | 9 | 1180 | |||
BC020987.1 | - | 4 | 496 | 3852258 | ||
BC027207.2 | - | 8 | 974 | |||
D83782.2 | - | 12 | 1761 |