Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ANKRD62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277333.1 | NP_001264262.1 | 13 | 2451 | ||
XM_017025720.1 | XP_016881209.1 | 13 | 2504 | |||
GenBank mRNA | BX649021.1 | - | 8 | 1546 |
Gene Symbol: ANKRD62P1-PARP4P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040115.1 | 1 | 138 | |||
GenBank mRNA | DN918251.1 | - | 1 | 50 |