Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098844.2 | NP_001092314.1 | 4 | 901 | ||
XM_011519627.2 | XP_011517929.1 | 4 | 778 | |||
XM_011519628.1 | XP_011517930.1 | 3 | 619 | |||
XM_011519629.1 | XP_011517931.1 | 3 | 667 | |||
XM_017016574.1 | XP_016872063.1 | 4 | 1627 | |||
GenBank mRNA | AK300456.1 | - | 3 | 644 | ||
BC157856.1 | - | 4 | 901 | 9057075 |