Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bex4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212457.2 | NP_997622.1 | 3 | 589 | ||
GenBank mRNA | AK010400.1 | - | 3 | 592 | ||
AY833558.1 | - | 3 | 589 | |||
BC090663.1 | - | 3 | 566 | 30677421 | ||
BC116226.2 | - | 3 | 493 | 40091511 |