Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077447.2 | NP_001070915.1 | 3 | 370 | ||
NM_001287506.1 | NP_001274435.1 | 3 | 394 | |||
NM_001287507.1 | NP_001274436.1 | 3 | 611 | |||
NM_001287508.1 | NP_001274437.1 | 3 | 466 | |||
NM_001287509.1 | NP_001274438.1 | 3 | 325 | |||
NM_001287510.1 | NP_001274439.1 | 2 | 988 | |||
NM_024664.3 | NP_078940.2 | 3 | 815 | |||
GenBank mRNA | AK021900.1 | - | 3 | 750 | ||
BC062586.1 | - | 2 | 938 | |||
BC104938.1 | - | 3 | 815 | 8143941 | ||
BC106064.1 | - | 3 | 765 | 3913530 | ||
BC112015.1 | - | 3 | 815 | 8327483 | ||
BM923950.1 | - | 3 | 618 | |||
BP211065.1 | - | 3 | 321 | |||
CB998485.1 | - | 3 | 278 | |||
DB479832.1 | - | 3 | 458 |