Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025099.5 | NP_079375.3 | 13 | 2284 | ||
NR_046431.1 | 13 | 2179 | ||||
XM_006721577.3 | XP_006721640.1 | 12 | 2139 | |||
XM_006721578.3 | XP_006721641.1 | 13 | 2268 | |||
XM_011524010.2 | XP_011522312.1 | 13 | 2163 | |||
XM_011524011.2 | XP_011522313.1 | 12 | 2033 | |||
GenBank mRNA | AK025823.1 | - | 3 | 427 | ||
AK091077.1 | - | 13 | 2235 | |||
AK225375.1 | - | 3 | 427 | |||
AK310636.1 | - | 2 | 453 | |||
AL831955.1 | - | 13 | 2126 | |||
BC026057.2 | - | 4 | 425 | |||
BC110373.1 | - | 5 | 651 | |||
BC111783.1 | - | 13 | 2235 | 40054070 | ||
BX406692.2 | - | 1 | 37 | |||
DQ451688.1 | - | 13 | 2225 | |||
FJ790670.1 | - | 13 | 2225 |