Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC18B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052831.2 | NP_439896.1 | 14 | 1960 | ||
XM_017010236.1 | XP_016865725.1 | 14 | 2151 | |||
GenBank mRNA | AJ420537.1 | - | 8 | 1106 | ||
AK124442.1 | - | 14 | 1839 |