Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318043.1 | NP_001304972.1 | 2, 3 | 308 | ||
NM_080820.5 | NP_543010.3 | 2, 3 | 308 | |||
XM_005260872.4 | XP_005260929.1 | 2, 3 | 849 | |||
GenBank mRNA | AF332356.1 | - | 2, 3 | 176 | ||
AK074304.1 | - | 2, 3 | 238 | |||
AK291440.1 | - | 2, 3 | 215 | |||
BC045167.2 | - | 2, 3 | 270 | 5294744 | ||
BC100923.1 | - | 2, 3 | 229 | 40003912 | ||
BC100924.2 | - | 2, 3 | 229 | 40003913 | ||
BC100925.1 | - | 2, 3 | 229 | 40003914 | ||
BC100926.1 | - | 2, 3 | 229 | 40003915 | ||
BI465352.1 | - | 2, 3 | 299 |