Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000801.4 | NP_000792.1 | 3 | 317 | ||
NM_054014.3 | NP_463460.1 | 3 | 317 | |||
GenBank mRNA | AB241123.1 | - | 2 | 95 | ||
AB241125.1 | - | 4 | 225 | |||
BC001925.1 | - | 3 | 191 | 2823635 | ||
BC005147.2 | - | 3 | 174 | 3504715 | ||
BC119732.1 | - | 3 | 254 | 40118164 | ||
BT007066.1 | - | 3 | 143 | |||
CR407613.1 | - | 3 | 143 | |||
CR542168.1 | - | 3 | 143 | |||
CX870892.1 | - | 3 | 317 | |||
M34539.1 | - | 3 | 221 | |||
X52220.1 | - | 3 | 173 |