Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TYMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113755.2 | NP_001107227.1 | 3 | 542 | ||
NM_001113756.2 | NP_001107228.1 | 2 | 714 | |||
NM_001257988.1 | NP_001244917.1 | 3 | 565 | |||
NM_001257989.1 | NP_001244918.1 | 3 | 530 | |||
NM_001953.4 | NP_001944.1 | 3 | 530 | |||
GenBank mRNA | AK057214.1 | - | 2 | 626 | ||
AK225269.1 | - | 3 | 460 | |||
AK298691.1 | - | 2 | 614 | |||
AK301194.1 | - | 3 | 457 | |||
AK314716.1 | - | 3 | 469 | |||
BC018160.1 | - | 3 | 441 | 3879593 | ||
BC052211.2 | - | 3 | 479 | 5174407 | ||
KU178037.1 | - | 2 | 338 | |||
KU178038.1 | - | 2 | 338 | |||
M63193.1 | - | 2 | 461 |