Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006273.3 | NP_006264.2 | 2 | 206 | ||
GenBank mRNA | AF043338.1 | - | 1 | 62 | ||
BC092436.1 | - | 2 | 200 | 30418073 | ||
BC112258.1 | - | 2 | 196 | 8327726 | ||
BC112260.1 | - | 2 | 196 | 8327728 | ||
CD523198.1 | - | 2 | 209 | |||
X71087.1 | - | 2 | 201 |