Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029872.1 | NP_084148.1 | 1 | 1645 | ||
GenBank mRNA | AK172180.1 | - | 1 | 159 | ||
BC023150.1 | - | 1 | 477 |