- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 5 | 657 | ||
NM_001271900.1 | NP_001258829.1 | 6 | 851 | |||
NM_001271948.1 | NP_001258877.1 | 5 | 1184 | |||
NM_181674.2 | NP_858060.2 | 5 | 982 | |||
NM_181675.3 | NP_858061.2 | 5 | 704 | |||
NM_181676.2 | NP_858062.1 | 4 | 788 | |||
NM_181677.2 | NP_858063.1 | 6 | 614 | |||
NM_181678.2 | NP_858064.1 | 6 | 627 | |||
NR_073526.1 | 4 | 1099 | ||||
NR_073527.1 | 5 | 736 | ||||
GenBank mRNA | AK056192.1 | - | 5 | 664 | ||
AK289717.1 | - | 5 | 633 | |||
AK294659.1 | - | 6 | 760 | |||
AK295347.1 | - | 5 | 628 | |||
AK314810.1 | - | 5 | 981 | |||
AL540315.3 | - | 3 | 317 | |||
BC031790.1 | - | 5 | 872 | 4939981 | ||
BI669304.1 | - | 6 | 612 | |||
BX647887.1 | - | 5 | 717 | |||
M64930.1 | - | 4 | 910 |