Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023096.2 | NP_075584.1 | 2 | 739 | ||
XM_008764847.1 | XP_008763069.1 | 2 | 1459 | |||
GenBank mRNA | AF196965.1 | - | 2 | 550 | ||
AF385401.1 | - | 2 | 739 |