Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134432.2 | NP_602308.1 | 2, 3 | 1037 | ||
XM_008772597.1 | XP_008770819.1 | 3, 4 | 1237 | |||
GenBank mRNA | BC078741.1 | - | 2, 3 | 1037 | 7128542 | |
BC087679.1 | - | 1, 2 | 873 | 7318795 |