Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Notch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008714.3 | NP_032740.3 | 3, 4 | 675 | ||
GenBank mRNA | AF508809.1 | - | 3, 4 | 490 | ||
AK090118.1 | - | 3, 4 | 651 | |||
BC138441.1 | - | 3, 4 | 490 | 8861461 | ||
BC138442.1 | - | 3, 4 | 490 | 8861462 | ||
Z11886.1 | - | 3, 4 | 489 |