Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Avpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011924.2 | NP_036054.1 | 1 | 977 | ||
GenBank mRNA | AK050523.1 | - | 1 | 59 |
Set Membership: |
![]() |