Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008238.3 | NP_001008239.1 | 8 | 1002 | ||
NM_016787.4 | NP_058067.2 | 8 | 1002 | |||
GenBank mRNA | AF035207.1 | - | 8 | 854 | ||
AF400107.1 | - | 7 | 742 | |||
AK014659.1 | - | 8 | 954 | |||
AK054288.1 | - | 8 | 968 | |||
AK159745.1 | - | 8 | 965 | |||
AK161116.1 | - | 8 | 955 | |||
AK169609.1 | - | 8 | 987 | |||
AY591757.1 | - | 7 | 721 | |||
AY591758.1 | - | 7 | 721 | |||
BC012670.1 | - | 8 | 961 | 4037627 |