Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135211.2 | NP_001128683.1 | 6 | 846 | ||
NM_001281304.1 | NP_001268233.1 | 5 | 847 | |||
NM_003602.4 | NP_003593.3 | 6 | 937 | |||
XM_006716153.2 | XP_006716216.1 | 5 | 721 | |||
XM_017012741.1 | XP_016868230.1 | 6 | 826 | |||
XM_017012742.1 | XP_016868231.1 | 6 | 844 | |||
GenBank mRNA | AF038847.1 | - | 6 | 801 | ||
AF447060.1 | - | 5 | 804 | |||
AK302121.1 | - | 6 | 834 | |||
AK310255.1 | - | 7 | 1109 | |||
BC036817.1 | - | 6 | 809 | 5744865 |