Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019968.2 | NP_064352.2 | 3 | 507 | ||
GenBank mRNA | AF205936.1 | - | 3 | 455 | ||
AK154684.1 | - | 1 | 3791 | |||
AK205970.1 | - | 2 | 111 | |||
BC115498.2 | - | 3 | 448 | 40100921 |